Order Entry
Singapore
ContactUsLinkComponent
 
 
 
Lithium hydroxide monohydrate ≥98.0% ACS, Fluka™
  HONC402974-500G
 :  
 
 
Lithium hydroxide monohydrate ≥98.0% ACS, Fluka™
  HONC402974-500G
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

 

 

  • Assay:
    Min. 98.0 %
  • Assay of Li₂CO₃:
    Max. 2.0 %
  • Water insoluble matter:
    Max. 0.01 %
  • Iron (Fe):
    Max. 0.002 %
  • Heavy metals (ICP) ACS:
    Max. 0.002 %
  • Chloride (Cl):
    Max. 0.01 %
  • Sulfate (SO₄):
    Max. 0.05 %

 

 

 :
 :41.96 g/mol
 :13,05556
 :920 °C (1013 hPa)
 :423 °C
 :1,830 g/cm³ (20 °C)
 :Ambient
 :215-183-4
 :2680
 :8,II
 :GHS07
 :GHS05
 :1310-66-3
 :LiOH·1H₂O
 :MFCD00149772