Order Entry
Singapore
ContactUsLinkComponent
Potassium hydroxide 0.5 mol/l (0.5 N) in methanol, AVS TITRINORM volumetric solution
Potassium hydroxide 0.5 mol/l (0.5 N) in methanol, AVS TITRINORM volumetric solution
Catalog # VWRC31984.293
Supplier:  VWR Chemicals
CAS Number:  
undefined
Potassium hydroxide 0.5 mol/l (0.5 N) in methanol, AVS TITRINORM volumetric solution
Catalog # VWRC31984.293
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

Specification Test Results

  • NIST traceable:
    Confirmed
  • Titer (20°C; real value 0.2 % accuracy):
    0.499 - 0.501 mol/l

Specification Test Results

About this item

Synonyms: Potash caustic, Caustic potash
 :
 :56.11 g/mol
Storage Temperature:Ambient
EINECS:215-181-3
UN:2924
ADR:3 (8),II
 :GHS02
 :GHS06
 :GHS08
 :GHS05
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553