Polysorbate 40
1
Catalog # EDQMY0000290
Supplier: EDQM
Jump to:
Product Details & Documents
Synonyms: Sorbitan monopalmitate, ethoxylated, Polyoxyethylene sorbitan monopalmitate
- CAS Number:9005-66-7
- Formula:C₆₂H₁₂₂O₂₆
- MDL Number:MFCD00165345
- Storage Temperature:Ambient
Specifications
Specifications
- Pk:1



