Order Entry
Singapore
ContactUsLinkComponent
Tween® 20 (Polysorbate) for molecular biology
Tween® 20 (Polysorbate) for molecular biology
Supplier:  PanReac AppliChem
Certificates
Tween® 20 (Polysorbate) for molecular biology
Supplier:  PanReac AppliChem
Certificates
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • DNases/RNases/Proteases:
    not detectable
  • Density (d 20°C/4°C):
    1.095 - 1.105
  • Hydroxyl-No.:
    approx. 100
  • Saponification No.:
    40 - 50