Order Entry
Singapore
ContactUsLinkComponent
Bovine serum albumin (BSA)
Bovine serum albumin (BSA)
Catalog # APLIA0850.0050
Supplier:  PanReac AppliChem
CAS Number:  
Bovine serum albumin (BSA)
Catalog # APLIA0850.0050
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • CAS No.:
    9048-46-8
    90604-29-8
  • Source:
    Serum
  • Species:
    Bovine
  • Storage conditions:
    Refrigerator
  • Protein synonyms:
    BSA|Fraction V|Cohn Fraction V
  • Protein/peptide name:
    Bovine Serum Albumin
  • Colour:
    light yellow
  • Grade:
    for radioimmunoassay (RIA)/EIA
  • Tested applications:
    Radioimmunoassay (RIA)/EIA

Specifications

About this item