Chicken native albumin (from egg)
Catalog # APLIA4344.1000
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- CAS No.:9006-59-1
- Protein/peptide type:Native
- Source:Egg
- Species:Chicken
- Storage conditions:Refrigerator
- Protein/peptide name:Albumin