Order Entry
Singapore
ContactUsLinkComponent
Human albumin (from serum), MP Biomedicals
Human albumin (from serum), MP Biomedicals
Supplier:  MP Biomedicals
Certificates
Human albumin (from serum), MP Biomedicals
Supplier:  MP Biomedicals

Specifications

  • CAS No.:
    70024-90-7
  • Source:
    Serum
  • Species:
    Human
  • Storage conditions:
    4 °C
  • Protein synonyms:
    Human Serum Albumin|HSA|serum albumin|HAS|serum of albumin
  • Protein/peptide name:
    Albumin
  • Purity:
    >97%
  • Molecular weight:
    65 kDa
  • Colour:
    Off-white
  • Formulation:
    Crystal like powder

Specifications