


Mercury(II) acetate 6% (w/v) in glacial acetic acid USP Test Solution (TS)
Catalog # RICCR4629000-1C
Supplier: Ricca Chemical
CAS Number:
Specifications
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:318.68 g/mol
Merck Index:13,05898
Melting Pt:178…180 °C
Density:3,27 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:216-491-1
UN:1629
ADR:6.1,II
:GHS06
:GHS08
:GHS09
CAS Number:1600-27-7
Formula:(H₃CCOO)₂Hg
MDL Number:MFCD00012165