Lactic acid 90% ≥90% in aqueous solution, TECHNICAL
Supplier: VWR Chemicals
About this item
Synonyms: DL-Lactic acid, 2-Hydroxypropionic acid, 2-Hydroxypropanoic acid
- CAS Number:50-21-5
- Formula:H₃CCH(OH)COOH
- MDL Number:MFCD00004520
- Molecular Weight:90.08 g/mol
- Boiling Pt:122 °C (20 hPa)
- Melting Pt:53 °C
- Density:1,11…1.21 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-018-0
2 Options Available Below
Product Details & Documents
Synonyms: DL-Lactic acid, 2-Hydroxypropionic acid, 2-Hydroxypropanoic acid
- CAS Number:50-21-5
- Formula:H₃CCH(OH)COOH
- MDL Number:MFCD00004520
- Molecular Weight:90.08 g/mol
- Boiling Pt:122 °C (20 hPa)
- Melting Pt:53 °C
- Density:1,11…1.21 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-018-0
Documents
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 25L Plastic drum for liquids DIN 61 (ZPACK13952500)
Packaging Information
P20_VWRC20366.293_SPEC_EN.pdf P20_VWRC20366.464_SPEC_EN.pdf spec_VWRC20366.293 spec_VWRC20366.293 spec_VWRC20366.464
Product specification
VWR Chemicals Big Packs and Bulks
Product Literature
Specifications for Product Family
Specification Test Results
- Assay:Min. 90 %



