D-(-)-Fructose
Glass bottle for solids, 100 mg
Catalog # EDQMF0550000
Supplier: EDQM
Jump to:
Product Details & Documents
Synonyms: D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose
- CAS Number:57-48-7
- Formula:C₆H₁₂O₆
- MDL Number:MFCD00148910
- Molecular Weight:180.16 g/mol
- Merck Index:13,04295
- Boiling Pt:552 °C (1013 hPa)
- Melting Pt:100…110 °C
- Density:1,59 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-333-3
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg



