



Petroleum spirit, 100…120 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:64742-49-0
:64742-82-1
MDL Number:MFCD00081849
Boiling Pt:100…120 °C (1013 hPa)
Density:0,7 g/cm³ (20 °C)
Flash Pt:–10 °C
Storage Temperature:Ambient
EINECS:265-151-9
UN:1268
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
Specification Test Results
- Substances coloured by H₂SO₄:Passes test
- Acidity:Max. 0.0003 meq/g
- Density (20/4):0.690 - 0.750
- Distillation (min. 90 %):100 - 125 °C
- Evaporation residue:Max. 20 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 225L Metal drum UN (ZPACE8010050) 30L Metal drum for liquids UN X (ZPACK12124125)
Packaging Information
spec_VWRC23845.323 spec_VWRC23845.323 spec_VWRC23845.323 spec_VWRC23845.460 spec_VWRC23845.460
Product specification
More
VWR Chemicals Big Packs and Bulks