



Petroleum spirit, 100…120 °C, AnalaR NORMAPUR® ACS analytical reagent
VWRC101814R
:
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
- Acidity:Max. 0.00005 meq/g
- Aromatics (as C₆H₆):Max. 20 ppm
- Bromine value:Max. 3.0
- Colouration:Max. 10 APHA
- Distillation (min. 90 %):100 -- 120 °C
- Evaporation residue:Max. 10 ppm
- Substances coloured by H₂SO₄:Max. 10 APHA
- Sulphur compounds (as S):Max. 50 ppm
- Water:Max. 100 ppm
- Al (Aluminium):Max. 0.1 ppm
- Ba (Barium):Max. 0.05 ppm
- Ca (Calcium):Max. 0.5 ppm
- Cd (Cadmium):Max. 0.05 ppm
- Co (Cobalt):Max. 0.02 ppm
- Cr (Chromium):Max. 0.02 ppm
- Cu (Copper):Max. 0.02 ppm
- Fe (Iron):Max. 0.1 ppm
- K (Potassium):Max. 0.2 ppm
- Mg (Magnesium):Max. 0.05 ppm
- Mn (Manganese):Max. 0.02 ppm
- Na (Sodium):Max. 0.5 ppm
- Ni (Nickel):Max. 0.02 ppm
- Pb (Lead):Max. 0.05 ppm
- Sn (Tin):Max. 0.1 ppm
- Sr (Strontium):Max. 0.02 ppm
- Zn (Zinc):Max. 0.1 ppm
- Conforms to ACS:Passes test
:
:100…120 °C (1013 hPa)
:0,7 g/cm³ (20 °C)
:–10 °C
:Ambient
:265-151-9
:1268
:3,II
:GHS02
:GHS08
:GHS07
:GHS09
:64742-49-0
:64742-82-1
:MFCD00081849