tert-Butyl-4-methoxyphenol (mixture of isomers)
Catalog # EDQMB1212000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:180.25 g/mol
Merck Index:12,01582
Boiling Pt:264 °C (1013 hPa)
Melting Pt:48…55 °C
Density:1,05 g/cm³ (20 °C)
Flash Pt:156 °C
Storage Temperature:Ambient
EINECS:246-563-8
CAS Number:25013-16-5
Formula:C₁₁H₁₆O₂
MDL Number:MFCD01779059