Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
Tantalum
Tantalum ≥99.95% (metals basis), thin foil, Thickness: 0.0127 mm (0.0005 in)
Tantalum ≥99.95% (metals basis), thin foil, Thickness: 0.0127 mm (0.0005 in)
:
Thermo Fisher Scientific
Tantalum ≥99.95% (metals basis), thin foil, Thickness: 0.0127 mm (0.0005 in)
:
Thermo Fisher Scientific
:
7440-25-7
:
:
Ta
:
MFCD00011252
:
180.95 g/mol
:
12,09223
:
5427 °C (1013 hPa)
:
2996 °C
:
16,69 g/cm³ (20 °C)
:
Ambient
:
231-135-5
Environmentally Preferable:
msds_ALFA014266
Thermo Scientific Acroseal 25 Years