Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Gold
Gold ≥99.9975% (metals basis), foil, Premion®, Thickness: 0.1 mm (0.004 in)
Gold ≥99.9975% (metals basis), foil, Premion®, Thickness: 0.1 mm (0.004 in)
:
Thermo Fisher Scientific
Gold ≥99.9975% (metals basis), foil, Premion®, Thickness: 0.1 mm (0.004 in)
:
Thermo Fisher Scientific
:
7440-57-5
:
:
Au
:
MFCD00003436
:
196.97 g/mol
:
12,04526
:
2800 °C (1013 hPa)
:
1063 °C
:
19,3 g/cm³ (20 °C)
:
Ambient
:
231-165-9
msds_ALFA011391
Thermo Scientific Acroseal 25 Years
Thermo Scientific Gold Products