Gold ≥99.9985% (metals basis), foil, Premion®, Thickness: 0.5 mm (0.02 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-57-5
:
Formula:Au
MDL Number:MFCD00003436
:196.97 g/mol
Merck Index:12,04526
Boiling Pt:2800 °C (1013 hPa)
Melting Pt:1063 °C
Density:19,3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-165-9