Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tin
Tin ≥98.8% (metals basis), foil, Thickness: 0.05 mm (0.002 in)
Tin ≥98.8% (metals basis), foil, Thickness: 0.05 mm (0.002 in)
:
Thermo Fisher Scientific
Tin ≥98.8% (metals basis), foil, Thickness: 0.05 mm (0.002 in)
:
Thermo Fisher Scientific
:
7440-31-5
:
:
Sn
:
MFCD00133862
:
12,09587
:
2270 °C (1013 hPa)
:
232 °C
:
7,31 g/cm³ (25 °C)
:
118.71 g/mol
:
Ambient
:
231-141-8
msds_ALFA041604