Tin ≥99.9985% (metals basis), wire, Puratronic®, Ø 0.5 mm (0.02 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-31-5
:
Formula:Sn
MDL Number:MFCD00133862
Merck Index:12,09587
Boiling Pt:2270 °C (1013 hPa)
Melting Pt:232 °C
Density:7,31 g/cm³ (25 °C)
:118.71 g/mol
Storage Temperature:Ambient
EINECS:23-141-8