Silicon ≥99.9999% (metals basis), polycrystalline rods, unpolished, Ø 1.6cm (0.6 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-21-3
:
Formula:Si
MDL Number:MFCD00085311
Density:2.34 g/cm³ (20 °C)
Molecular Weight:28.09 g/mol
Merck Index:12,08635
Storage Temperature:Ambient
EINECS:231-130-8
Specifications
- Environmentally Preferable: