Germanium ≥99.999% (metals basis), pieces 3-9 mm (0.1-0.4 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-56-4
:
Formula:Ge
MDL Number:MFCD00085310
Boiling Pt:2830
Melting Pt:937.2
Density:5.323 g/cm³ (20 °C)
Molecular Weight:72.61 g/mol
Merck Index:12,04415
Storage Temperature:Ambient
EINECS:231-164-3
Specifications
- Environmentally Preferable: