Order Entry
Singapore
ContactUsLinkComponent
Gallium ≥99.99999% (metals basis), pellets, Ø 6 mm
Gallium ≥99.99999% (metals basis), pellets, Ø 6 mm
Catalog # ALFA44325.18
CAS Number:  
Gallium ≥99.99999% (metals basis), pellets, Ø 6 mm
Catalog # ALFA44325.18
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368
  • Environmentally Preferable:

Specifications

About this item

CAS Number:7440-55-3
 :
Formula:Ga
MDL Number:MFCD00134045
 :69.72 g/mol
Merck Index:12,04364
Boiling Pt:2403 °C (1013 hPa)
Melting Pt:29 °C
Density:5,904 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:231-163-8
UN:2803
ADR:8,III
 :GHS05