Aluminium ≥99.1% (metals basis), thin foil, Thickness: 2.0 µm (0.00008 in)
Catalog # ALFA40708.GH
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:1736
- Pk Info:100×100 mm
Specifications
About this item
CAS Number:7429-90-5
:
Formula:Al
MDL Number:MFCD00134029
:26.98 g/mol
Merck Index:13,00321
Boiling Pt:2327 °C (1013 hPa)
Melting Pt:660 °C
Density:2,7 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-072-3