VWR® IPASEPT® 70, γ-Irradiated
Supplier: VWR Collection
Certificates
About this item
70% Iso-propyl alcohol compliant with Ph. Eur. and 30% water.
Filtered through a 0.2 µm filter, packaged under nitrogen.
70% Iso-propyl alcohol compliant with Ph. Eur. and 30% water.
Filtered through a 0.2 µm filter, packaged under nitrogen.