Order Entry
Singapore
ContactUsLinkComponent
VWR® Magnetised pipette rack
  VWRU82007-220
undefined
VWR® Magnetised pipette rack
  VWRU82007-220
  • Colour:
    Blue
  • Description:
    Magnetised pipette rack
  • W×D×H:
    85×290×410 mm
  • Pk:
    1
This durable blue ABS plastic storage rack features circular magnets for secure mounting to metal fume hoods, refrigerators, file cabinets, and more.
  • Four angled compartments accommodate 0,1 to 25 ml pipettes
  • Includes two movable attachments to hold pipetting devices