Zinc sulphate heptahydrate, J.T.Baker®
BAKR4382.1000
: Avantor
:
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
- Assay:99.0 - 103.0 %
- Ammonium (NH4):max. 0.001 %
- Calcium (Ca):max. 0.005 %
- Potassium (K):max. 0.01 %
- Magnesium (Mg):max. 0.005 %
- Sodium (Na):max. 0.05 %
- Lead (Pb):max. 0.003 %
- Insoluble Matter:max. 0.01 %
- Nitrate (NO3):max. 0.002 %
- pH of 5% Solution at 25°C:4.4 - 6.0
- Chloride (Cl):max. 5
- Iron (Fe):max. 5
- Manganese (Mn):max. 3
:
:9,III
:287.54 g/mol
:3077
:7446-20-0
:ZnSO₄·7H₂O
:MFCD00149894