Organic reference standard, Xylazine hydrochloride, Ph.Eur., RS
Catalog # EDQMX0075000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:125 mg
- Standards:Reference standard
- CAS No.:23076-35-9
- Description:Xylazine hydrochloride
- Formula:C₁₂H₁₇ClN₂S
- Storage temperature:Ambient
- Description:Xylazine hydrochloride
Specifications
About this item
Filled under inert atmosphere protected from humidity.
- In Monograph 1481
- Solubility in water: 100 - 1000 g/l