Organic reference standard, Warfarin sodium, Ph.Eur., RS
Catalog # EDQMW0100000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:10 mg
- Standards:Reference standard
- CAS No.:129-06-6
- Description:Warfarin sodium
- Formula:C₁₉H₁₅NaH₄
- Storage temperature:Ambient
- Description:Warfarin sodium
Specifications
About this item
Filled under inert atmosphere protected from humidity.
- In Monograph 0698
- Solubility in water: more than 1000 g/l