Organic reference standard, Vinblastine sulphate, Ph.Eur., RS
Catalog # EDQMV0300000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:5 mg
- Standards:Reference standard
- CAS No.:143-67-9
- Description:Vinblastine sulphate
- Formula:C₄₆H₆₀N₄O₁₃S
- Storage temperature:-80°C ± 10°C
- Description:Vinblastine sulphate
Specifications
About this item
- In Monographs 0748 and 0749
- Solubility in water: 100 - 1000 g/l