Organic reference standard, Verapamil impurity M, Ph.Eur., RS
EDQMV0100026
: EDQM
:
- Pack type:Vial
- Pk:10 mg
- Standards:Reference standard
- Description:Verapamil impurity M
- Formula:C₄₂H₅₇N₃O₆
- Storage temperature:Ambient
- Description:Verapamil impurity M
- In Monograph 0573
- Solubility in water: less than 0,1 g/l