Organic reference standard, Tylosin, Ph.Eur., RS
Catalog # EDQMT2880000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:150 mg
- Standards:Reference standard
- CAS No.:1401-69-0
- Description:Tylosin
- Formula:C₄₆H₇₇NO₁₇
- Storage temperature:Ambient
- Description:Tylosin
Specifications
About this item
- In Monographs 1273, 1274, 1661, and 20702
- Partial solubility in water
- Pharmaceutical applications