Organic reference standard, Trimyristin, Ph.Eur., RS
Catalog # EDQMT2500100
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:500 mg
- Standards:Reference standard
- CAS No.:555-45-3
- Description:Trimyristin
- Formula:C₄₅H₈₆O₆
- Storage temperature:Ambient
- Description:Trimyristin
Specifications
About this item
- In Monograph 1410
- Pharmaceutical applications