Order Entry
Singapore
ContactUsLinkComponent
Organic reference standard, Trimetazidine for system suitability, Ph.Eur., RS
Organic reference standard, Trimetazidine for system suitability, Ph.Eur., RS
Catalog # EDQMY0000093
Supplier:  EDQM
CAS Number:  
Organic reference standard, Trimetazidine for system suitability, Ph.Eur., RS
Catalog # EDQMY0000093
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    50 mg
  • Standards:
    Reference standard
  • CAS No.:
    13171-25-0
  • Description:
    Trimetazidine for system suitability
  • Formula:
    C₁₄H₂₄Cl₂N₂O₃
  • Storage temperature:
    Ambient
  • Description:
    Trimetazidine for system suitability

Specifications

About this item

  • In Monograph 1741
  • Soluble in/Miscible with methanol
  • Pharmaceutical applications