Organic reference standard, Tricaprylin, Ph.Eur., RS
Catalog # EDQMT1978000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:225 mg
- Standards:Reference standard
- CAS No.:538-23-8
- Description:Tricaprylin
- Formula:C₂₇H₅₀O₆
- Storage temperature:Ambient
- Description:Tricaprylin
Specifications
About this item
- In Monograph 1410
- Pharmaceutical applications