Organic reference standard, Tricaprin, Ph.Eur., RS
Catalog # EDQMT1977990
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:150 mg
- Standards:Reference standard
- CAS No.:621-71-6
- Description:Tricaprin
- Formula:C₃₃H₆₂O₆
- Storage temperature:Ambient
- Description:Tricaprin
Specifications
About this item
- In Monograph 1410
- Pharmaceutical applications