Order Entry
Singapore
ContactUsLinkComponent
Organic reference standards, Tribenoside, Ph.Eur., RS
Organic reference standards, Tribenoside, Ph.Eur., RS
Catalog # EDQMY0000166
Supplier:  EDQM
CAS Number:  
Organic reference standards, Tribenoside, Ph.Eur., RS
Catalog # EDQMY0000166
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    70 mg
  • Standards:
    Reference standard
  • CAS No.:
    53928-30-6
  • Description:
    Tribenoside impurity A
  • Formula:
    C₃₀H₃₄O₆
  • Storage temperature:
    15 to 25°C
  • Description:
    Tribenoside impurity A

Specifications

About this item

  • In Monograph 1740
  • Pharmaceutical applications