Organic reference standard, trans-Epoxyphytomenadione, Ph.Eur., RS
Catalog # EDQMP1609010
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:10 mg
- Standards:Reference standard
- CAS No.:85955-78-8
- Description:trans-Epoxyphytomenadione
- Environmentally Preferable:
- Formula:C₃₁H₄₆O₃
- Description:trans-Epoxyphytomenadione
Specifications
About this item
- In Monograph 1036