Organic reference standard, Tioconazole for system suitability, Ph.Eur., RS
Catalog # EDQMY0000282
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:50 mg
- Standards:Reference standard
- CAS No.:65899-73-2
- Description:Tioconazole for system suitability
- Formula:C₁₆H₁₃Cl₃N₂OS
- Storage temperature:Ambient
- Description:Tioconazole for system suitability
Specifications
About this item
- In Monograph 2074
- Solubility in water: 0,1 to 1 g/l