Organic reference standard, Tiabendazole, Ph.Eur., RS
Catalog # EDQMT1410000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:50 mg
- Standards:Reference standard
- CAS No.:148-79-8
- Description:Tiabendazole
- Formula:C₁₀H₇N₃S
- Storage temperature:Ambient
- Description:Tiabendazole
Specifications
About this item
- In Monograph 0866
- Solubility in water: less than 0,1 g/l