Organic reference standard, Testosterone for system suitability, Ph.Eur., RS
Catalog # EDQMY0000343
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:20 mg
- Standards:Reference standard
- CAS No.:58-22-0
- Description:Testosterone for system suitability
- Formula:C₁₉H₂₈O₂
- Storage temperature:Ambient
- Description:Testosterone for system suitability
Specifications
About this item
- In Monograph 1373
- Soluble in/miscible with alcohol and mthylene chloride