Organic reference standard, Terconazole, Ph.Eur., RS
Catalog # EDQMT0060000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:250 mg
- Standards:Reference standard
- CAS No.:67915-31-5
- Description:Terconazole
- Formula:C₂₆H₃₁Cl₂N₅O₃
- Storage temperature:Ambient
- Description:Terconazole
Specifications
About this item
- In Monograph 1270
- Solubility in water: less than 0,1 g/l