Organic reference standard, Terbutaline sulfate, Ph.Eur., RS
Catalog # EDQMT0050000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:100 mg
- Standards:Reference standard
- CAS No.:23031-32-5
- Description:Terbutaline sulfate
- Formula:C₂₄H₄₀N₂O₁₀S
- Storage temperature:Ambient
- Description:Terbutaline sulfate
Specifications
About this item
- In Monographs 0690 and 2451
- Soluble in water
- Pharmaceutical applications