Organic reference standards, Sulpiride, Ph.Eur., RS
Catalog # EDQMS2191000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:20 mg
- Standards:Reference standard
- CAS No.:26116-12-1
- Description:Sulpiride impurity A
- Formula:C₇H₁₆N₂
- Storage temperature:Ambient
- Description:Sulpiride impurity A
Specifications
About this item
- Sulpiride In Monographs 1045 and 1348
- Sulpiride impurity A in Monographs 1045 and 1490
- Pharmaceutical applications