Organic reference standard, Sulfapyridine, Ph.Eur., RS
Catalog # EDQMS2159100
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:20 mg
- Standards:Reference standard
- CAS No.:144-82-1
- Description:Sulfapyridine
- Formula:C₁₁H₁₁N₃O₂S
- Storage temperature:Ambient
- Description:Sulfapyridine
Specifications
About this item
- In Monograph 0863
- Soluble in/miscible with dimethyl sulfoxide and methanol
- Pharmaceutical applications