Organic reference standard, Sulfaguanidine, Ph.Eur., RS
Catalog # EDQMS1975600
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:60 mg
- Standards:Reference standard
- CAS No.:57-67-0
- Description:Sulfaguanidine
- Formula:C₇H₁₀N₄O₂S
- Storage temperature:Ambient
- Description:Sulfaguanidine
Specifications
About this item
- In Monograph 0295 and 1476
- Pharmaceutical applications