Order Entry
Singapore
ContactUsLinkComponent
 
Organic reference standard, Sulfadoxine, Ph.Eur., RS
Catalog # EDQMS1950000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Sulfadoxine, Ph.Eur., RS
Catalog # EDQMS1950000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    50 mg
  • Standards:
    Reference standard
  • CAS No.:
    2447-57-6
  • Description:
    Sulfadoxine
  • Formula:
    C₁₂H₁₄N₄O₄S
  • Storage temperature:
    Ambient
  • Description:
    Sulfadoxine

Specifications

About this item

  • In Monograph 0740
  • Soluble in/miscible with DMSO and methanol
  • Pharmaceutical applications