Organic reference standards, Sotalol, Ph.Eur., RS
Catalog # EDQMY0000114
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:25 mg
- Standards:Reference standard
- CAS No.:959-24-0
- Description:Sotalol hydrochloride
- Formula:C₁₂H₂₁ClN₂O₃S
- Storage temperature:Ambient
- Description:Sotalol hydrochloride
Specifications
About this item
- In Monograph 2004
- Solubility in water: 100 - 1000 g/l