Organic reference standard, Somatropin, Ph.Eur., RS
Catalog # EDQMS0947000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:1
- Standards:Reference standard
- CAS No.:12629-01-5
- Description:Somatropin
- Formula:C₉₉₀H₁₅₂₉N₂₆₃O₂₉₉S₇
- Storage temperature:-20°C
- Description:Somatropin
Specifications
About this item
- In Monographs 0950, 0951, 0952, and 2370