Organic reference standard, Sodium taurocholate BRP, Ph.Eur., RS
Catalog # EDQMS0900000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:10.000 mg
- Standards:Reference standard
- CAS No.:145-42-6
- Description:Sodium taurocholate BRP
- Formula:C₂₆H₄₄NNaO₇S
- Storage temperature:Ambient
- Description:Sodium taurocholate BRP
Specifications
About this item
- In Monograph 0350
- Soluble in water
- Pharmaceutical applications