Order Entry
Singapore
ContactUsLinkComponent
Organic reference standard, Sodium hyaluronate BRP, Ph.Eur., RS
Organic reference standard, Sodium hyaluronate BRP, Ph.Eur., RS
Catalog # EDQMS0780000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Sodium hyaluronate BRP, Ph.Eur., RS
Catalog # EDQMS0780000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    300 mg
  • Standards:
    Reference standard
  • CAS No.:
    9067-32-7
  • Description:
    Sodium hyaluronate BRP
  • Formula:
    C₁₄H₂₂NNaO₁₁
  • Storage temperature:
    Ambient
  • Description:
    Sodium hyaluronate BRP

Specifications

About this item

  • In Monograph 0912
  • Pharmaceutical applications