Order Entry
Singapore
ContactUsLinkComponent
 
Organic reference standard, Sodium cromoglicate, Ph.Eur., RS
  EDQMS0750000
 :  EDQM
 :  
Organic reference standard, Sodium cromoglicate, Ph.Eur., RS
  EDQMS0750000
 :  EDQM
 :  

 

  • Pack type:
    Vial
  • Pk:
    10 mg
  • Standards:
    Reference standard
  • CAS No.:
    15826-37-6
  • Description:
    Sodium cromoglicate
  • Formula:
    C₂₃H₁₄Na₂O₁₁
  • Storage temperature:
    Ambient
  • Description:
    Sodium cromoglicate

 

 

  • In Monograph 0562
  • Pharmaceutical applications